COMH4211F0EC Combi-Blocks, Inc.

1-(4-Fluorobenzoyl)-4-phenylbuta-1,3-diene

  • MDL number MFCD28657104
  • Cas Number : 99646-84-1
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7639
Availability details for
Quantity
  • Availability
  • Add to Cart