COMH4211F0FA Combi-Blocks, Inc.

Adamantane-1-carboxamidine hydrochloride

  • MDL number MFCD00167793
  • Cas Number : 50417-14-6
  • Molecular Weight : 214.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7742
Availability details for
Quantity
  • Availability
  • Add to Cart