COMH4211F0FC Combi-Blocks, Inc.

2-(2-Dimethylaminoethyl)-5-methyl-2h-pyrazole-3-ylamine

  • MDL number MFCD03789157
  • Cas Number : 685892-22-2
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7753
Availability details for
Quantity
  • Availability
  • Add to Cart