COMH4211F102 Combi-Blocks, Inc.

(1-(3-Chlorophenyl)cyclobutyl)methanamine

  • MDL number MFCD09904043
  • Cas Number : 115816-34-7
  • Molecular Weight : 195.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7834
Availability details for
Quantity
  • Availability
  • Add to Cart