COMH4211F111 Combi-Blocks, Inc.

(+/-)-Jasmonic acid

  • MDL number MFCD04221274
  • Cas Number : 221682-41-3
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-7956
Availability details for
Quantity
  • Availability
  • Add to Cart