COMH4211F177 Combi-Blocks, Inc.

(3-Fluorobicyclo[1.1.1]pentan-1-yl)methanol

  • MDL number MFCD29037345
  • Cas Number : 262852-01-7
  • Molecular Weight : 116.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-8776

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-8776
Availability details for
Quantity
  • Availability
  • Add to Cart