COMH4211F1B9 Combi-Blocks, Inc.

3-Aminocyclobutanecarboxylic acid

  • MDL number MFCD09864182
  • Cas Number : 160191-58-2
  • Molecular Weight : 115.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9274
Availability details for
Quantity
  • Availability
  • Add to Cart