COMH4211F1F6 Combi-Blocks, Inc.

4-Chloro-7-cyclohexyl-6,7-dihydro-2-(methylthio)-(5h)-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD00202983
  • Cas Number : 388572-67-6
  • Molecular Weight : 283.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9740
Availability details for
Quantity
  • Availability
  • Add to Cart