COMH4211F215 Combi-Blocks, Inc.

5-Methylthiophene-2-sulfonamide

  • MDL number MFCD08444612
  • Cas Number : 53595-69-0
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0182

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0182
Availability details for
Quantity
  • Availability
  • Add to Cart