COMH4211F218 Combi-Blocks, Inc.

3-(2-Methyl-1h-indol-3-yl)-acrylic acid ethyl ester

  • MDL number MFCD01415789
  • Cas Number : 854694-25-0
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0254
Availability details for
Quantity
  • Availability
  • Add to Cart