COMH4211F23E Combi-Blocks, Inc.

2-Amino-1-naphthoic acid

  • MDL number MFCD17012404
  • Cas Number : 79979-69-4
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0948
Availability details for
Quantity
  • Availability
  • Add to Cart