COMH4211F24D Combi-Blocks, Inc.

1-(4-Thien-2-ylphenyl)ethanone

  • MDL number MFCD04039158
  • Cas Number : 107834-03-7
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1155

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1155
Availability details for
Quantity
  • Availability
  • Add to Cart