COMH4211F25E Combi-Blocks, Inc.

(S)-4-Benzyl-3-pentanoyloxazolidin-2-one

  • MDL number MFCD20269711
  • Cas Number : 143868-89-7
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1470
Availability details for
Quantity
  • Availability
  • Add to Cart