COMH4211F2C1 Combi-Blocks, Inc.

1,4,5,6-Tetrahydro-1-methylpyrrolo-[3,4-c]-pyrazole dihydrochloride

  • MDL number MFCD16613983
  • Cas Number : 1187830-68-7
  • Molecular Weight : 196.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-3581
Availability details for
Quantity
  • Availability
  • Add to Cart