COMH4211F38E Combi-Blocks, Inc.

1,4-Diazabicyclo[3.2.1]octane

  • MDL number MFCD19216438
  • Cas Number : 5167-08-8
  • Molecular Weight : 112.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8501

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8501
Availability details for
Quantity
  • Availability
  • Add to Cart