COMH4211F391 Combi-Blocks, Inc.

N-(P-Tolylsulfonylmethyl)formamide

  • MDL number MFCD00015915
  • Cas Number : 36635-56-0
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8545
Availability details for
Quantity
  • Availability
  • Add to Cart