COMH4211F3B0 Combi-Blocks, Inc.

5-Methyl-1-phenylpyrazole-4-carbonyl chloride

  • MDL number MFCD02677692
  • Cas Number : 205113-77-5
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9131

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-9131
Availability details for
Quantity
  • Availability
  • Add to Cart