COMH4211F3FC Combi-Blocks, Inc.

N-Methoxy-n-methylcyclopentanecarboxamide

  • MDL number MFCD12546236
  • Cas Number : 167303-65-3
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2195
Availability details for
Quantity
  • Availability
  • Add to Cart