COMH4211F3FE Combi-Blocks, Inc.

2-(4-Bromo-2-methoxyphenoxy)ethanol

  • MDL number MFCD17676517
  • Cas Number : 877399-29-6
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-2373
Availability details for
Quantity
  • Availability
  • Add to Cart