COMH4211F404 Combi-Blocks, Inc.

5-(Chloromethyl)-4-methyl-1,3-thiazole

  • MDL number MFCD08060637
  • Cas Number : 10014-52-5
  • Molecular Weight : 147.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-3483
Availability details for
Quantity
  • Availability
  • Add to Cart