COMH4211F40C Combi-Blocks, Inc.

(3S)-3-(([(tert-Butoxy)carbonyl]amino)methyl)-5-methylhexanoic acid

  • MDL number MFCD21247143
  • Cas Number : 649748-09-4
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4859

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-4859
Availability details for
Quantity
  • Availability
  • Add to Cart