COMH4211F427 Combi-Blocks, Inc.

6-Ethynylimidazo[1,2-a]pyridine

  • MDL number MFCD23703386
  • Cas Number : 1373337-86-0
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-6516

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-6516
Availability details for
Quantity
  • Availability
  • Add to Cart