COMH4211F430 Combi-Blocks, Inc.

3-Amino-4-bromo-2,6-dichlorobenzoic acid

  • MDL number MFCD25542117
  • Cas Number : 1415124-69-4
  • Molecular Weight : 284.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7076
Availability details for
Quantity
  • Availability
  • Add to Cart