COMH4211F438 Combi-Blocks, Inc.

6-Chloro-3-(2-furyl)[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD11182543
  • Molecular Weight : 220.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7263

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7263
Availability details for
Quantity
  • Availability
  • Add to Cart