COMH4211F43A Combi-Blocks, Inc.

6-Chloro-3-cyclopentyl-[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD11182546
  • Cas Number : 1094260-24-8
  • Molecular Weight : 222.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7267

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7267
Availability details for
Quantity
  • Availability
  • Add to Cart