COMH4211F43D Combi-Blocks, Inc.

Ethyl 2-amino-4-chloro-5h-pyrrolo[3,4-d]pyrimidine-6(7h)-carboxylate

  • MDL number MFCD14706243
  • Cas Number : 1046861-17-9
  • Molecular Weight : 242.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7402
Availability details for
Quantity
  • Availability
  • Add to Cart