COMH4211F466 Combi-Blocks, Inc.

Ethyl 3-(2-chlorophenyl)-1h-pyrazole-5-carboxylate

  • MDL number MFCD09749412
  • Molecular Weight : 250.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-9407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-9407
Availability details for
Quantity
  • Availability
  • Add to Cart