COMH4211F46D Combi-Blocks, Inc.

1-(2-Fluoro-5-(trifluoromethyl)phenyl)ethanamine

  • MDL number MFCD18702359
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-9832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-9832
Availability details for
Quantity
  • Availability
  • Add to Cart