COMH4211F474 Combi-Blocks, Inc.

1,3-dimethyl 4-fluorobenzene-1,3-dicarboxylate

  • MDL number MFCD22490724
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4765
Availability details for
Quantity
  • Availability
  • Add to Cart