COMH4211F476 Combi-Blocks, Inc.

5-Bromo-N,2-dimethylpyridine-3-carboxamide

  • MDL number MFCD32065770
  • Cas Number : 2365418-34-2
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4792
Availability details for
Quantity
  • Availability
  • Add to Cart