COMH4211F478 Combi-Blocks, Inc.

3,4-Dihydro-1H-1,7-naphthyridin-2-one

  • MDL number MFCD08234987
  • Cas Number : 885272-20-8
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4806
Availability details for
Quantity
  • Availability
  • Add to Cart