COMH4211F47C Combi-Blocks, Inc.

2-(4-Iodophenyl)-N-methylacetamide

  • MDL number MFCD29965342
  • Cas Number : 355117-64-5
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4823

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4823
Availability details for
Quantity
  • Availability
  • Add to Cart