COMH4211F47F Combi-Blocks, Inc.

N-ethyl-6-methoxypyridine-2-carboxamide

  • Cas Number : 1863037-84-6
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4833
Availability details for
Quantity
  • Availability
  • Add to Cart