COMH4211F484 Combi-Blocks, Inc.

4-[(6-Methoxypyridin-2-yl)carbonyl]morpholine

  • MDL number MFCD32065778
  • Cas Number : 1791419-73-2
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4838
Availability details for
Quantity
  • Availability
  • Add to Cart