COMH4211F487 Combi-Blocks, Inc.

N-Methyl-2-(4-nitrophenoxy)acetamide

  • MDL number MFCD05344680
  • Cas Number : 20916-14-7
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4849

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4849
Availability details for
Quantity
  • Availability
  • Add to Cart