COMH4211F48C Combi-Blocks, Inc.

tert-Butyl 4-[2-(4-nitrophenoxy)acetyl]piperazine-1-carboxylate

  • MDL number MFCD09149287
  • Cas Number : 927604-76-0
  • Molecular Weight : 365.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4854
Availability details for
Quantity
  • Availability
  • Add to Cart