COMH4211F48D Combi-Blocks, Inc.

2-(4-Nitrophenoxy)-1-(piperidin-1-yl)ethanone

  • MDL number MFCD00548559
  • Cas Number : 50508-35-5
  • Molecular Weight : 264.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4855
Availability details for
Quantity
  • Availability
  • Add to Cart