COMH4211F494 Combi-Blocks, Inc.

tert-Butyl 4-(3-bromo-4-chlorophenoxy)piperidine-1-carboxylate

  • MDL number MFCD32065786
  • Cas Number : 2253767-82-5
  • Molecular Weight : 390.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4908
Availability details for
Quantity
  • Availability
  • Add to Cart