COMH4211F49C Combi-Blocks, Inc.

4-(6-Fluoropyridin-2-yl)morpholine

  • Cas Number : 1248137-75-8
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4934

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4934
Availability details for
Quantity
  • Availability
  • Add to Cart