COMH4211F49D Combi-Blocks, Inc.

N,2-Dimethyl-5-nitrobenzenesulfonamide

  • Cas Number : 126148-12-7
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4952
Availability details for
Quantity
  • Availability
  • Add to Cart