COMH42122323 Combi-Blocks, Inc.

6-Nitrocinnolin-3-ol

  • MDL number MFCD32065789
  • Cas Number : 2365418-43-3
  • Molecular Weight : 191.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2199

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2199
Availability details for
Quantity
  • Availability
  • Add to Cart