COMH42122325 Combi-Blocks, Inc.

tert-Butyl 6-(trifluoromethyl)indole-1-carboxylate

  • MDL number MFCD32065794
  • Cas Number : 2365418-87-5
  • Molecular Weight : 285.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7046
Availability details for
Quantity
  • Availability
  • Add to Cart