COMH42122326 Combi-Blocks, Inc.

2-Chloro-4-(dihydroxyboranyl)-6-fluorobenzoic acid

  • Molecular Weight : 218.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7224
Availability details for
Quantity
  • Availability
  • Add to Cart