COMH42122334 Combi-Blocks, Inc.

Methyl 2-hydroxy-2'-methyl-[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD27991971
  • Cas Number : 1141474-11-4
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-0629

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-0629
Availability details for
Quantity
  • Availability
  • Add to Cart