COMH42122340 Combi-Blocks, Inc.

(1-[4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenyl]cyclopropylmethanol

  • MDL number MFCD28133740
  • Cas Number : 1467060-42-9
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1217
Availability details for
Quantity
  • Availability
  • Add to Cart