COMH42122344 Combi-Blocks, Inc.

Methyl 3-(cyclopropylmethoxy)-4-iodobenzoate

  • MDL number MFCD28133757
  • Molecular Weight : 332.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1256

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1256
Availability details for
Quantity
  • Availability
  • Add to Cart