COMH42122346 Combi-Blocks, Inc.

tert-Butyl 4-(3-(4-bromophenoxy)propyl)piperazine-1-carboxylate

  • MDL number MFCD28133783
  • Cas Number : 1467060-86-1
  • Molecular Weight : 399.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1319
Availability details for
Quantity
  • Availability
  • Add to Cart