COMH42122348 Combi-Blocks, Inc.

1-(3-(4-Bromophenoxy)propyl)-4-methylpiperazine

  • MDL number MFCD02108511
  • Cas Number : 93699-37-7
  • Molecular Weight : 313.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-1321
Availability details for
Quantity
  • Availability
  • Add to Cart