COMH4212236F Combi-Blocks, Inc.

2-Amino-5-bromo-n-methoxy-n-methylbenzamide

  • MDL number MFCD12913689
  • Cas Number : 304854-54-4
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-3038
Availability details for
Quantity
  • Availability
  • Add to Cart