COMH42122384 Combi-Blocks, Inc.

2-(1-Methyl-1h-imidazol-4-yl)ethan-1-amine

  • MDL number MFCD01941538
  • Cas Number : 501-75-7
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-6553
Availability details for
Quantity
  • Availability
  • Add to Cart